6-Fluoro-1H-indole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A159641-100MG
A159641-10G
A159641-250MG
A159641-5G
103805-272EA
31
USD
103805-272
103843-494
77011-154
77011-156
77011-158
77300-410
6-Fluoro-1H-indole-3-carboxylic acid 98%
6-Fluoro-1H-indole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₆FNO₂ MW: 179.148 g/mol Boiling Pt: 422.2 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD06203359 CAS Number: 23077-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...