6-Fluoroindole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-1H-indole

A301354-25G A301354-5G A301354-100G A301354-10G A301354-500G
103835-454EA 8 USD
103835-454 76838-142 76838-144 77277-326 77277-328 77277-330
6-Fluoroindole 98%
6-Fluoroindole

Precaución: for Research Use Only

Formula: C₈H₆FN
Boiling Pt: 258 °C (760 mm Hg)
Melting Pt: 72…77 °C
Flash Pt: 109.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR