6-Fluoro-1H-indazole 97%
Supplier: Ambeed
|
Warning
|
A140889-10G
A140889-25G
A140889-5G
A140889-1G
103802-916EA
24
USD
103802-916
77162-304
77299-168
77299-170
77316-034
6-Fluoro-1H-indazole 97%
6-Fluoro-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₅FN₂ MW: 136.1264 g/mol Boiling Pt: 274.9 °C (760 mm Hg) Flash Pt: 120.051 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07371562 CAS Number: 348-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...