6-Carboxyfluorescein 98%

Supplier: Ambeed
Warning

Synonyms: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

A135997-100MG A135997-10G A135997-1MG A135997-250MG A135997-25G A135997-1G A135997-5G
103799-354EA 24 USD
103799-354 103802-258 76767-494 76767-496 76767-498 76767-500 76767-502 77315-138 77315-140
6-Carboxyfluorescein 98%
6-Carboxyfluorescein

Precaución: for Research Use Only

Formula: C₂₁H₁₂O₇
MW: 376.32 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036873
CAS Number: 3301-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR