6-Ethynyl-4,4-dimethylthiochroman 98%

Supplier: Ambeed
Warning

A299771-1G A299771-100MG
103814-246EA 40 USD
103814-246 103835-396 76838-084 77114-448
6-Ethynyl-4,4-dimethylthiochroman 98%
6-Ethynyl-4,4-dimethylthiochroman

Precaución: for Research Use Only

Formula: C₁₃H₁₄S
MW: 202.32 g/mol
Boiling Pt: 299.2 °C (760 mm Hg)
Melting Pt: 69…72 °C
Flash Pt: 128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00902029
CAS Number: 118292-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR