6-Ethylpyridin-3-amine 98%

Supplier: Ambeed
Danger

A597757-100MG A597757-250MG A597757-5G
103828-966EA 174 USD
103828-966 76819-378 76819-380 76819-382
6-Ethylpyridin-3-amine 98%
6-Ethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
Boiling Pt: 244 °C (760 mm Hg)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD11111131
CAS Number: 126553-00-2
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR