6-Cyclopropylpyridin-3-amine 95%

Supplier: Ambeed
Warning

A196996-100MG A196996-250MG A196996-1G A196996-5G
103816-884EA 137 USD
103816-884 103847-498 76814-118 76814-120 77085-476 77085-478
6-Cyclopropylpyridin-3-amine 95%
6-Cyclopropylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD12033370
CAS Number: 1159821-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR