6-Cyclopropyl-2,3-dihydropyridazin-3-one 97%

Supplier: Ambeed
Warning

A321837-1G A321837-250MG
103825-294EA 53 USD
103825-294 103836-228 76839-142 77890-643
6-Cyclopropyl-2,3-dihydropyridazin-3-one 97%
6-Cyclopropyl-2,3-dihydropyridazin-3-one

Precaución: for Research Use Only

Formula: C₇H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD16871334
CAS Number: 1161737-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR