6-Cyanopicolinic acid 98%

Supplier: Ambeed
Warning

A148851-100MG A148851-250MG
103803-972EA 97 USD
103803-972 76973-002 76973-004
6-Cyanopicolinic acid 98%
6-Cyanopicolinic acid

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10574696
CAS Number: 872602-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR