6-Cyanonicotinic acid 97%

Supplier: Ambeed
Warning

A122730-10G A122730-250MG A122730-1G A122730-5G
103800-266EA 63 USD
103800-266 76766-632 76766-634 77297-888 77297-890
6-Cyanonicotinic acid 97%
6-Cyanonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 395.9 °C (760 mm Hg)
Melting Pt: 190…194 °C
Flash Pt: 193.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01318135
CAS Number: 70165-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR