Indole-6-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 6-Cyano-1H-indole, 6-Cyanoindole, 1H-Indole-6-carbonitrile

A267680-25G A267680-5G A267680-100G A267680-10G A267680-500G A267680-1G
103812-874EA 11 USD
103812-874 103852-774 77092-520 77092-522 77262-368 77262-370 77262-372 77304-986
Indole-6-carbonitrile 98%
Indole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MW: 142.16 g/mol
Boiling Pt: 350 °C (760 mm Hg)
Melting Pt: 130…132 °C
Flash Pt: 121.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00016732
CAS Number: 15861-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR