6-Cyano-1-tetralone 98%

Supplier: Ambeed
Warning

A218732-10G A218732-250MG A218732-1G
103818-736EA 88 USD
103818-736 103849-606 76800-834 76800-836 77088-132
6-Cyano-1-tetralone 98%
6-Cyano-1-tetralone

Precaución: for Research Use Only

Formula: C₁₁H₉NO
Boiling Pt: 345.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02179289
CAS Number: 90401-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR