6-Chloroquinoline 98%

Supplier: Ambeed
Warning

A181298-25G A181298-100G A181298-10G A181298-1G A181298-5G
103845-962EA 10 USD
103845-962 77083-488 77260-252 77260-254 77260-256 77301-714
6-Chloroquinoline 98%
6-Chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 263 °C (760 mm Hg)
Melting Pt: 41…43 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00024024
CAS Number: 612-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR