6-Chloro-4-pyrimidinecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloropyrimidine-4-carboxylic acid

A113206-250MG A113206-1G A113206-25G A113206-100MG
103798-188EA 99 USD
103798-188 103824-026 76766-012 77310-850 77570-728 77623-938
6-Chloro-4-pyrimidinecarboxylic acid 97%
6-Chloro-4-pyrimidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Boiling Pt: 343.69 °C (760 mm Hg)
Flash Pt: 161.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08692928
CAS Number: 37131-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR