6-Chloropyrimidine-4-carbonitrile 98%

Supplier: Ambeed
Warning

A337001-10G A337001-1G
103825-734EA 125 USD
103825-734 76817-226 76839-712
6-Chloropyrimidine-4-carbonitrile 98%
6-Chloropyrimidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂ClN₃
MW: 139.54
Melting Pt: 25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607731
CAS Number: 939986-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR