6-Chloropyrimidin-4(1H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-4(1H)-pyrimidinone, 6-Chloro-4-pyrimidinol, 4-Chloro-6-hydroxypyrimidine

A343446-1G A343446-10G
103825-886EA 9 USD
103825-886 103836-862 76839-908 77290-562 77290-564
6-Chloropyrimidin-4(1H)-one 97%
6-Chloropyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₄H₃ClN₂O
Boiling Pt: 199.4 °C (760 mm Hg)
Melting Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00233957
CAS Number: 4765-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR