6-Chloronicotinamide 97%
Supplier: Ambeed
|
Warning
|
A137113-100G
A137113-10G
A137113-25G
103802-410EA
6
USD
103802-410
77095-260
77095-262
77298-888
77315-362
6-Chloronicotinamide 97%
6-Chloronicotinamide
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O Boiling Pt: 331 °C (760 mm Hg) Melting Pt: 210…212 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006242 CAS Number: 6271-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...