6-Chloropyrazine-2-carbonitrile 97%

Supplier: Ambeed
Warning

A294501-1G A294501-25G A294501-5G A294501-250MG
103814-010EA 14 USD
103814-010 103835-150 76837-786 76837-788 76837-790 77141-396
6-Chloropyrazine-2-carbonitrile 97%
6-Chloropyrazine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂ClN₃
MW: 139.5400 g/mol
Boiling Pt: 229.3 °C (760 mm Hg)
Melting Pt: 24…26 °C
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607699
CAS Number: 6863-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR