6-Chloro-N-methylpyrimidin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A127168-100G
A127168-10G
A127168-250MG
A127168-1G
A127168-25G
A127168-5G
103800-924EA
9
USD
103800-924
77108-956
77108-958
77108-960
77313-162
77313-164
77313-166
6-Chloro-N-methylpyrimidin-4-amine 97%
6-Chloro-N-methylpyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₅H₆ClN₃ MW: 143.5700 g/mol Boiling Pt: 279.7 °C (760 mm Hg) Melting Pt: 138…142 °C Flash Pt: 122.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09702146 CAS Number: 65766-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...