6-Chloro-n,n-dimethyl-3-pyridinamine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-N,N-dimethylpyridin-3-amine

A177362-250MG A177362-1G
103807-228EA 125 USD
103807-228 103845-556 76985-656 77301-502
6-Chloro-n,n-dimethyl-3-pyridinamine 98%
6-Chloro-n,n-dimethyl-3-pyridinamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.613 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00234337
CAS Number: 41288-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR