6-Chloro-2,3-dihydro-1H-isoindol-1-one 96%

Supplier: Ambeed
Warning

Synonyms: 6-Chloroisoindolin-1-one

A287437-250MG A287437-1G A287437-5G
103813-752EA 38 USD
103813-752 103834-894 76772-412 76837-444 76837-446
6-Chloro-2,3-dihydro-1H-isoindol-1-one 96%
6-Chloro-2,3-dihydro-1H-isoindol-1-one

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.592 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234734
CAS Number: 58083-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR