6-Chloroimidazo[1,2-b]pyridazin-3-amine 97%

Supplier: Ambeed
Warning

A407834-10G A407834-250MG A407834-25G A407834-5G
103827-164EA 23 USD
103827-164 103838-206 76773-774 76773-776 76773-778 76841-702
6-Chloroimidazo[1,2-b]pyridazin-3-amine 97%
6-Chloroimidazo[1,2-b]pyridazin-3-amine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD09881451
CAS Number: 166176-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR