6-Chloroimidazo[1,2-a]pyrazine 95%

Supplier: Ambeed
Warning

A316777-100MG A316777-1G A316777-25G A316777-5G A316777-10G
103825-118EA 33 USD
103825-118 103836-048 76772-860 76838-888 76838-890 76838-892 77626-580
6-Chloroimidazo[1,2-a]pyrazine 95%
6-Chloroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Melting Pt: 137…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909644
CAS Number: 76537-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR