6-Chloro-1-benzothiophene-2-carboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: 6-Chlorobenzo[b]thiophene-2-carboxylic acid

A433573-100MG A433573-250MG
103827-556EA 31 USD
103827-556 103838-624 76773-946 76773-948
6-Chloro-1-benzothiophene-2-carboxylic acid 96%
6-Chloro-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅ClO₂S
MW: 212.66 g/mol
Boiling Pt: 408.6 °C (760 mm Hg)
Melting Pt: 271…274 °C at N/A mmHg
Flash Pt: 200.9 °C
Storage Temperature: Freezer
MDL Number: MFCD07377059
CAS Number: 26018-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR