6-Chloro-9-methylpurine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-9-methyl-9H-purine, NSC 4948

A106414-100MG A106414-1G A106414-250MG
103797-034EA 133 USD
103797-034 103822-788 76765-496 77283-478 77592-134
6-Chloro-9-methylpurine 97%
6-Chloro-9-methylpurine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
MW: 168.59 g/mol
Boiling Pt: 321.7 °C (760 mm Hg)
Melting Pt: 129…134 °C (760 mm Hg)
Flash Pt: 148.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00127840
CAS Number: 2346-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR