6-Chloro-9-[2,3-O-(1-methylethylidene)-beta-D-ribofuranosyl]-9H-Purine 97%
Supplier: Ambeed
|
Warning
|
A310639-10G
A310639-250MG
A310639-1G
A310639-5G
103814-628EA
15
USD
103814-628
103835-812
77002-680
77002-682
77306-030
77306-032
6-Chloro-9-[2,3-O-(1-methylethylidene)-beta-D-ribofuranosyl]-9H-Purine 97%
6-Chloro-9-[2,3-O-(1-methylethylidene)-beta-D-ribofuranosyl]-9H-Purine
Precaución: for Research Use Only
Formula:
C₁₃H₁₅ClN₄O₄ Boiling Pt: 527.8 °C (760 mm Hg) Melting Pt: 156…159 °C Flash Pt: 272.996 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788833 CAS Number: 39824-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...