6-Chloro-9-(tetrahydro-2H-pyran-2-yl)-9H-purine 99%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-9-(tetrahydro-2-pyranyl)purine, 6-Chloro-9-(tetrahydropyran-2-yl)purine, 6-Chloro-9-(2-tetrahydropyranyl)purine

A250440-10G A250440-1G A250440-5G A250440-100G A250440-500G
103811-926EA 5 USD
103811-926 103851-766 76815-642 77091-120 77304-530 77612-416 77672-232
6-Chloro-9-(tetrahydro-2H-pyran-2-yl)-9H-purine 99%
6-Chloro-9-(tetrahydro-2H-pyran-2-yl)-9H-purine

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₄O
MW: 238.6735 g/mol
Boiling Pt: 428.3 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00022821
CAS Number: 7306-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR