6-Chloro-7-methyl-7H-purine ≥96%

Supplier: Ambeed
Warning

A146025-100MG A146025-10G A146025-250MG A146025-1G
103803-620EA 77 USD
103803-620 103841-706 76767-992 76767-994 76767-996 77317-012
6-Chloro-7-methyl-7H-purine ≥96%
6-Chloro-7-methyl-7H-purine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
Boiling Pt: 321.7 °C (760 mm Hg)
Melting Pt: 175…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01943043
CAS Number: 5440-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR