6-Chloro-5-nitropicolinic acid 98%

Supplier: Ambeed
Warning

A185986-10G A185986-250MG
103815-900EA 35 USD
103815-900 103846-474 76985-906 77683-474
6-Chloro-5-nitropicolinic acid 98%
6-Chloro-5-nitropicolinic acid

Precaución: For research use only.

Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01927937
CAS Number: 353277-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR