2-Chloro-6-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-methyl-5-nitropyridine, 6-Chloro-5-nitro-2-picoline, 2-Chloro-3-nitro-6-picoline, 2-Chloro-3-nitro-6-methylpyridine

A167132-100G A167132-10G A167132-250MG A167132-500G A167132-1G A167132-25G A167132-5G
103806-132EA 6 USD
103806-132 103844-394 77096-350 77096-352 77096-354 77096-356 77320-680 77320-682 77320-684
2-Chloro-6-methyl-3-nitropyridine 98%
2-Chloro-6-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57
Boiling Pt: 281.7 °C (760 mm Hg)
Melting Pt: 70…74 °C
Flash Pt: 124.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03085820
CAS Number: 56057-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR