6-Chloro-5-methylpyridazin-3-amine 98%

Supplier: Ambeed
Warning

A153266-100G A153266-10G A153266-250MG A153266-1G A153266-25G A153266-5G
103804-480EA 16 USD
103804-480 103842-652 76871-726 76871-728 76871-730 77318-292 77318-294 77318-296
6-Chloro-5-methylpyridazin-3-amine 98%
6-Chloro-5-methylpyridazin-3-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.574 g/mol
Boiling Pt: 358.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD19203420
CAS Number: 66346-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR