6-Chloro-5-iodopyridin-2-amine 96%

Supplier: Ambeed
Warning

A795953-100MG A795953-250MG A795953-1G A795953-5G A795953-10G
103841-038EA 86 USD
103841-038 76832-276 76832-278 76845-284 76845-286 77886-772
6-Chloro-5-iodopyridin-2-amine 96%
6-Chloro-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
Boiling Pt: 340.4±42.0 °C (760 mm Hg)
Flash Pt: 159.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20486991
CAS Number: 1221398-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR