6-Chloro-5-fluoropyrimidin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A178447-100MG
A178447-10G
A178447-250MG
A178447-1G
A178447-25G
A178447-5G
103807-332EA
8
USD
103807-332
103845-672
76974-332
76974-334
76974-336
77083-102
77083-104
77083-106
6-Chloro-5-fluoropyrimidin-4-amine 97%
6-Chloro-5-fluoropyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₄H₃ClFN₃ Boiling Pt: 349.5 °C (760 mm Hg) Melting Pt: 112…115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD14706712 CAS Number: 851984-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...