6-Chloro-5-fluoronicotinonitrile 99%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-fluoropyridine-3-carbonitrile, 6-Chloro-3-cyano-5-fluoropyridine

A138296-100MG A138296-250MG A138296-1G A138296-5G
103802-594EA 63 USD
103802-594 76871-056 76871-058 77315-610 77315-612
6-Chloro-5-fluoronicotinonitrile 99%
6-Chloro-5-fluoronicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂ClFN₂
MW: 156.545 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09972184
CAS Number: 1020253-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR