6-Chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-(trifluoromethyl)nicotinic acid

A321637-100MG A321637-250MG A321637-1G A321637-5G
103825-284EA 117 USD
103825-284 103836-216 76828-216 76828-218 76839-126 76839-128
6-Chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid 97%
6-Chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11848376
CAS Number: 1110782-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR