6-Chloro-4-methylpyridine-2-carboxylic acid 98%

Supplier: Ambeed
Danger

A120219-10G A120219-5G A120219-1G
103799-900EA 18 USD
103799-900 76766-414 77297-744 77312-004
6-Chloro-4-methylpyridine-2-carboxylic acid 98%
6-Chloro-4-methylpyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 341.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09834827
CAS Number: 324028-95-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR