3-Amino-6-chloro-4-methylpyridine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-Amino-6-chloro-4-picoline, 5-Amino-2-chloro-4-methylpyridine, 6-Chloro-4-methylpyridin-3-amine
A268256-1G
A268256-25G
A268256-5G
A268256-100G
A268256-10G
103812-910EA
5
USD
103812-910
103852-812
77092-560
77092-562
77092-564
77113-860
77685-766
3-Amino-6-chloro-4-methylpyridine 97%
3-Amino-6-chloro-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂ Boiling Pt: 309.7 °C (760 mm Hg) Melting Pt: 69…73 °C Flash Pt: 141.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427661 CAS Number: 66909-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...