6-Chloro-4-methylpyridin-2-ol 97%

Supplier: Ambeed
Warning

A163233-100MG A163233-1G A163233-5G
103805-700EA 384 USD
103805-700 103843-958 77191-172 77300-644 77300-646
6-Chloro-4-methylpyridin-2-ol 97%
6-Chloro-4-methylpyridin-2-ol

Precaución: Inert atmosphere, room temperature

Formula: C₆H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD16610473
CAS Number: 25297-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR