6-Chloro-4-methylnicotinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-4-methyl-3-pyridinecarboxylic acid, 6-Chloro-4-methylpyridine-3-carboxylic acid, 5-Carboxy-2-chloro-4-methylpyridine, 2-Chloro-4-methylpyridine-5-carboxylic acid
A218917-100MG
A218917-250MG
A218917-25G
A218917-1G
A218917-5G
A218917-10G
103818-758EA
13
USD
103818-758
103849-632
76975-768
76975-770
76975-772
77088-174
77088-176
77671-954
6-Chloro-4-methylnicotinic acid 98%
6-Chloro-4-methylnicotinic acid
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.581 g/mol Boiling Pt: 344.7 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD12755871 CAS Number: 503555-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...