6-Chloro-4-methyl-3-nitropyridin-2-amine 95%

Supplier: Ambeed
Warning

A125150-100MG A125150-5G A125150-250MG
103800-630EA 61 USD
103800-630 76822-658 77298-036 77885-114
6-Chloro-4-methyl-3-nitropyridin-2-amine 95%

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O₂
Boiling Pt: 350.2±37.0 °C (760 mm Hg)
Flash Pt: 165.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17016063
CAS Number: 863878-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR