6-Chloro-4-methoxypyridin-3-amine 95%

Supplier: Ambeed
Warning

A330094-5G A330094-100MG A330094-250MG A330094-1G
103825-544EA 123 USD
103825-544 103836-502 76839-480 77194-988 77194-990 77306-406
6-Chloro-4-methoxypyridin-3-amine 95%
6-Chloro-4-methoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18257796
CAS Number: 1256805-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR