6-Chloro-4-methoxynicotinic acid 98%

Supplier: Ambeed
Warning

A161626-100MG A161626-10G A161626-250MG A161626-1G A161626-25G A161626-5G
103805-510EA 16 USD
103805-510 103843-748 76768-570 76768-572 76768-574 77300-542 77319-736 77319-738
6-Chloro-4-methoxynicotinic acid 98%
6-Chloro-4-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD10000638
CAS Number: 716362-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR