6-Chloropyrimidine-4,5-diamine 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Diamino-6-chloropyrimidine, 6-Chloro-4,5-diaminopyrimidine

A217917-1G A217917-25G A217917-5G A217917-10G A217917-250MG
103818-668EA 11 USD
103818-668 103849-398 77088-036 77088-038 77088-040 77112-656 77112-658
6-Chloropyrimidine-4,5-diamine 97%
6-Chloropyrimidine-4,5-diamine

Precaución: for Research Use Only

Formula: C₄H₅ClN₄
MW: 144.5600 g/mol
Boiling Pt: 336.7 °C (760 mm Hg)
Melting Pt: 238…239 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023270
CAS Number: 4316-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR