6-Chloro-4-(trifluoromethyl)nicotinic acid 95%

Supplier: Ambeed
Warning

A478533-100MG A478533-250MG A478533-1G
103828-100EA 58 USD
103828-100 103839-172 76774-332 76774-334 76843-110
6-Chloro-4-(trifluoromethyl)nicotinic acid 95%
6-Chloro-4-(trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
Boiling Pt: 309.9 °C (760 mm Hg)
Melting Pt: 95…105 °C
Flash Pt: 141.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06800446
CAS Number: 261635-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR