6-Chloro-3-nitroquinolin-4-ol 98%

Supplier: Ambeed
Warning

A149721-10G A149721-250MG A149721-5G
103804-074EA 33 USD
103804-074 103842-200 76768-114 76768-116 77317-702
6-Chloro-3-nitroquinolin-4-ol 98%
6-Chloro-3-nitroquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₃
MW: 224.6 g/mol
Boiling Pt: 368.5 °C (760 mm Hg)
Flash Pt: 176.7 °C
Storage Temperature: Ambient
MDL Number: MFCD09800425
CAS Number: 101861-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR