6-Chloro-2-hydroxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3-nitropyridin-2(1H)-one

A331747-1G A331747-10G A331747-250MG A331747-25G A331747-5G
103825-584EA 19 USD
103825-584 103836-550 76839-542 77263-600 77263-602 77306-416 77306-418
6-Chloro-2-hydroxy-3-nitropyridine 97%
6-Chloro-2-hydroxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
Boiling Pt: 313.2 °C (760 mm Hg)
Flash Pt: 143.2 °C
Storage Temperature: Ambient
MDL Number: MFCD05261002
CAS Number: 92138-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR