6-Chloro-3-nitropyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3-nitropicolinonitrile, 6-Chloro-2-cyano-3-nitropyridine

A130978-100G A130978-10G A130978-250MG A130978-1G A130978-25G A130978-5G
103801-508EA 19 USD
103801-508 76870-730 76870-732 76870-734 77298-442 77298-444 77313-986
6-Chloro-3-nitropyridine-2-carbonitrile 97%
6-Chloro-3-nitropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₂ClN₃O₂
MW: 183.552 g/mol
Boiling Pt: 347.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06657641
CAS Number: 93683-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR