6-Chloro-3-iodopyridin-2-amine 98%
Supplier: Ambeed
|
Warning
|
A414609-1G
A414609-25G
A414609-5G
A414609-10G
A414609-250MG
103827-278EA
15
USD
103827-278
103838-324
76841-890
76841-892
76841-894
77033-780
77033-782
6-Chloro-3-iodopyridin-2-amine 98%
6-Chloro-3-iodopyridin-2-amine
Precaución: for Research Use Only
Formula:
C₅H₄ClIN₂ Boiling Pt: 324.707 °C at N/A mmHg Flash Pt: 150.178 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13181515 CAS Number: 800402-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...