6-Chloro-3-iodoimidazo[1,2-a]pyridazine 96%

Supplier: Ambeed
Warning

A132051-100MG A132051-250MG A132051-1G A132051-5G
103801-682EA 48 USD
103801-682 76767-282 76767-284 77314-230 77314-232
6-Chloro-3-iodoimidazo[1,2-a]pyridazine 96%
6-Chloro-3-iodoimidazo[1,2-a]pyridazine

Precaución: for Research Use Only

Formula: C₆H₃ClIN₃
Storage Temperature: Refrigerator
MDL Number: MFCD11044749
CAS Number: 923595-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR