6-Chloro-3-iodo-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A130743-100MG A130743-250MG A130743-1G A130743-5G
103801-474EA 92 USD
103801-474 76983-680 76983-682 77298-430 77313-936
6-Chloro-3-iodo-1H-pyrrolo[3,2-c]pyridine 95%
6-Chloro-3-iodo-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClIN₂
MW: 278.47 g/mol
Boiling Pt: 415.4±40.0 °C (760 mm Hg)
Flash Pt: 205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09749980
CAS Number: 1000341-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR